4-[(1,1-Dimethylethyl)dimethylsilyl]-2-methyl-3-butyn-2-ol ≥95%

Supplier: AOBChem USA

Warning

386871-5G 386871-25G 386871-1G 386871-500MG
77629-024EA 1484.23 USD
77629-024 77635-756 77642-546 77646-582
4-[(1,1-Dimethylethyl)dimethylsilyl]-2-methyl-3-butyn-2-ol ≥95%
4-[(1,1-Dimethylethyl)dimethylsilyl]-2-methyl-3-butyn-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₂₂OSi
MW: 198.38 g/mol
Storage Temperature: Ambient
CAS Number: 911137-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR