2-Methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

38587-5G 38587-1G 38587-25G 38587-500MG
77634-962EA 322 USD
77634-962 77638-530 77659-118 77650-536
2-Methyl-2H-indazole ≥97%
2-Methyl-2H-indazole

Precaución: For Research Use Only

Formula: C₈H₈N₂
MW: 132.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234418
CAS Number: 4838-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR