5-Methyl-2-(4-nitrophenoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

385849-5G 385849-25G 385849-1G 385849-500MG
77629-018EA 1484.23 USD
77629-018 77635-752 77638-528 77650-534
5-Methyl-2-(4-nitrophenoxy)benzaldehyde ≥95%
5-Methyl-2-(4-nitrophenoxy)benzaldehyde

Precaución: For Research Use Only

Formula: 257.24 g/mol
MW: C₁₄H₁₁NO₄
Storage Temperature: Ambient
CAS Number: 1991821-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR