1-(5-Phenyl-2-thienyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

385156-5G 385156-1G 385156-500MG 385156-25G
77633-028EA 904 USD
77633-028 77638-526 77652-752 77655-536
1-(5-Phenyl-2-thienyl)-1-propanone ≥95%
1-(5-Phenyl-2-thienyl)-1-propanone

Precaución: For Research Use Only

Formula: 216.3 g/mol
MW: C₁₃H₁₂OS
Storage Temperature: Ambient
CAS Number: 1665-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR