5-Bromo-2-isopropylaniline ≥95%

Supplier: AOBChem USA

Warning

38435-5G 38435-25G 38435-1G 38435-500MG
77633-026EA 1333 USD
77633-026 77659-112 77642-536 77646-564
5-Bromo-2-isopropylaniline ≥95%
5-Bromo-2-isopropylaniline

Precaución: For Research Use Only

Formula: C₉H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD09221909
CAS Number: 923148-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR