1-Bromo-3-fluoro-2,4-diiodobenzene ≥95%

Supplier: AOBChem USA

Warning

384021-5G 384021-25G 384021-1G 384021-500MG
77629-012EA 1484.23 USD
77629-012 77655-534 77642-532 77646-562
1-Bromo-3-fluoro-2,4-diiodobenzene ≥95%
1-Bromo-3-fluoro-2,4-diiodobenzene

Precaución: For Research Use Only

Formula: 426.79 g/mol
MW: C₆H₂BrFI₂
Storage Temperature: Ambient
CAS Number: 1805538-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR