1-Bromo-2-fluoro-5-methoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

38389-5G 38389-1G 38389-500MG
77655-532EA 1988 USD
77655-532 77642-530 77650-528
1-Bromo-2-fluoro-5-methoxy-3-methylbenzene ≥95%
1-Bromo-2-fluoro-5-methoxy-3-methylbenzene

Precaución: For Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD28613072
CAS Number: 1782348-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR