1-[5-(1,1-Dimethylethyl)-2-methylphenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

383242-5G 383242-1G 383242-500MG
77659-108EA 2053 USD
77659-108 77642-528 77650-526
1-[5-(1,1-Dimethylethyl)-2-methylphenyl]-1-propanone ≥95%
1-[5-(1,1-Dimethylethyl)-2-methylphenyl]-1-propanone

Precaución: For Research Use Only

Formula: 204.31 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD14529920
CAS Number: 857391-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR