(3-Formyl-2,4-dimethoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

38271-5G 38271-1G 38271-500MG
77629-010EA 1484.23 USD
77629-010 77638-522 77646-558
(3-Formyl-2,4-dimethoxyphenyl)boronic acid ≥97%
(3-Formyl-2,4-dimethoxyphenyl)boronic acid

Precaución: For Research Use Only

Formula: C₉H₁₁BO₅
Storage Temperature: Ambient
MDL Number: MFCD32902640
CAS Number: 2375943-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR