1-(2-Methylnaphthalen-1-yl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

382396-5G 382396-1G 382396-25G 382396-500MG
77629-006EA 1484.23 USD
77629-006 77638-520 77659-106 77650-524
1-(2-Methylnaphthalen-1-yl)prop-2-en-1-ol ≥95%
1-(2-Methylnaphthalen-1-yl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 198.26 g/mol
MW: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD35199356
CAS Number: 1042012-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR