5-Iodoacenaphthene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Iodo-1,2-dihydroacenaphthylene, 5-Iodo-1,2-dihydroacenaphthylene, 1,2-Dihydro-5-iodoacenaphthylene

382325-5G 382325-500MG 382325-25G 382325-1G
77633-024EA 1055 USD
77633-024 77652-744 77655-528 77642-524
5-Iodoacenaphthene ≥95%
5-Iodoacenaphthene

Precaución: For Research Use Only

Formula: C₁₂H₉I
MW: 280.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00275570
CAS Number: 6861-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR