1-(1-Methyl-1H-imidazol-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

382311-5G 382311-1G 382311-25G 382311-500MG
77634-960EA 276 USD
77634-960 77638-516 77659-104 77650-522
1-(1-Methyl-1H-imidazol-2-yl)ethanone ≥95%
1-(1-Methyl-1H-imidazol-2-yl)ethanone

Precaución: For Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08668178
CAS Number: 85692-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR