(6-Bromo-2,3-difluorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

382098-5G 382098-1G 382098-25G 382098-500MG
77629-000EA 1484.23 USD
77629-000 77638-514 77659-100 77646-546
(6-Bromo-2,3-difluorophenyl)(phenyl)methanol ≥95%
(6-Bromo-2,3-difluorophenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 299.111 g/mol
MW: C₁₃H₉BrF₂O
Storage Temperature: Ambient
CAS Number: 3037703-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR