Cyclopropyl[2-methoxy-5-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

381259-5G 381259-25G 381259-1G 381259-500MG
77628-998EA 1484.23 USD
77628-998 77635-746 77642-520 77650-520
Cyclopropyl[2-methoxy-5-(trifluoromethyl)phenyl]methanone ≥95%
Cyclopropyl[2-methoxy-5-(trifluoromethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₁F₃O₂
Storage Temperature: Ambient
CAS Number: 1479016-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR