1-[2-Methoxy-5-(trifluoromethyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

381237-5G 381237-25G 381237-1G 381237-500MG
77628-996EA 1484.23 USD
77628-996 77659-098 77642-518 77646-542
1-[2-Methoxy-5-(trifluoromethyl)phenyl]-1-propanone ≥95%
1-[2-Methoxy-5-(trifluoromethyl)phenyl]-1-propanone

Precaución: For Research Use Only

Formula: 232.2 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
CAS Number: 1261877-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR