1-[2-Ethoxy-5-(trifluoromethyl)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

381236-5G 381236-1G 381236-500MG
77655-526EA 2281 USD
77655-526 77642-516 77650-518
1-[2-Ethoxy-5-(trifluoromethyl)phenyl]ethanone ≥95%
1-[2-Ethoxy-5-(trifluoromethyl)phenyl]ethanone

Precaución: For Research Use Only

Formula: 232.2 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
CAS Number: 1488557-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR