1-(3,5-Dichloro-2,6-difluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

380931-5G 380931-1G 380931-25G 380931-500MG
77628-992EA 1484.23 USD
77628-992 77638-510 77659-094 77646-538
1-(3,5-Dichloro-2,6-difluorophenyl)ethanol ≥95%
1-(3,5-Dichloro-2,6-difluorophenyl)ethanol

Precaución: For Research Use Only

Formula: 227.035 g/mol
MW: C₈H₆Cl₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD35199126
CAS Number: 3002554-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR