(3,5-Dichloro-2,6-difluorophenyl)(ethyl)sulphane ≥95%

Supplier: AOBChem USA

Warning

380927-5G 380927-1G 380927-500MG 380927-25G
77633-022EA 1441 USD
77633-022 77638-508 77652-742 77655-524
(3,5-Dichloro-2,6-difluorophenyl)(ethyl)sulphane ≥95%
(3,5-Dichloro-2,6-difluorophenyl)(ethyl)sulphane

Precaución: For Research Use Only

Formula: C₈H₆Cl₂F₂S
Storage Temperature: Ambient
CAS Number: 3037703-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR