2,6-Difluoro-3-(trimethylsilyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

380546-5G 380546-1G 380546-25G 380546-500MG
77628-990EA 1484.23 USD
77628-990 77638-506 77655-522 77646-536
2,6-Difluoro-3-(trimethylsilyl)benzonitrile ≥95%
2,6-Difluoro-3-(trimethylsilyl)benzonitrile

Precaución: For Research Use Only

Formula: 211.283 g/mol
MW: C₁₀H₁₁F₂NSi
Storage Temperature: Ambient
MDL Number: MFCD35198995
CAS Number: 3002450-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR