2,6-Difluoro-3-(2-hydroxypropan-2-yl)benzonitrile ≥95%
Supplier: AOBChem USA
|
Warning
|
380543-5G
380543-1G
380543-25G
380543-500MG
77628-988EA
1484.23
USD
77628-988
77638-504
77659-092
77646-534
2,6-Difluoro-3-(2-hydroxypropan-2-yl)benzonitrile ≥95%
2,6-Difluoro-3-(2-hydroxypropan-2-yl)benzonitrile
Precaución: For Research Use Only
Formula:
C₁₀H₉F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD35198992 CAS Number: 3002493-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...