3-(Cyclopropyl(hydroxy)methyl)-2,6-difluorobenzonitrile ≥95%
Supplier: AOBChem USA
|
Warning
|
380524-5G
380524-25G
380524-1G
380524-500MG
77628-986EA
1484.23
USD
77628-986
77655-520
77642-514
77646-532
3-(Cyclopropyl(hydroxy)methyl)-2,6-difluorobenzonitrile ≥95%
3-(Cyclopropyl(hydroxy)methyl)-2,6-difluorobenzonitrile
Precaución: For Research Use Only
Formula:
C₁₁H₉F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD35198974 CAS Number: 3002430-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...