2,6-Difluoro-3-propionylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

380512-5G 380512-25G 380512-1G 380512-500MG
77628-984EA 1484.23 USD
77628-984 77659-090 77642-512 77650-516
2,6-Difluoro-3-propionylbenzonitrile ≥95%
2,6-Difluoro-3-propionylbenzonitrile

Precaución: For Research Use Only

Formula: C₁₀H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD35198962
CAS Number: 3002446-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR