Ethyl 2-(2,4-difluoro-3-(methylthio)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

380497-5G 380497-1G 380497-500MG
77628-982EA 1484.23 USD
77628-982 77642-508 77646-530
Ethyl 2-(2,4-difluoro-3-(methylthio)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,4-difluoro-3-(methylthio)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 260.257 g/mol
MW: C₁₁H₁₀F₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD35198948
CAS Number: 1898380-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR