(2,4-Difluoro-3-(methylthio)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

380475-5G 380475-25G 380475-1G 380475-500MG
77633-020EA 1441 USD
77633-020 77659-088 77646-526 77646-528
(2,4-Difluoro-3-(methylthio)phenyl)(phenyl)methanone ≥95%
(2,4-Difluoro-3-(methylthio)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 264.29 g/mol
MW: C₁₄H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD35198926
CAS Number: 3002534-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR