Cyclobutyl(2,4-difluoro-3-(methylthio)phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

380472-5G 380472-25G 380472-1G 380472-500MG
77628-980EA 1484.23 USD
77628-980 77635-744 77638-502 77646-524
Cyclobutyl(2,4-difluoro-3-(methylthio)phenyl)methanone ≥95%
Cyclobutyl(2,4-difluoro-3-(methylthio)phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₂F₂OS
Storage Temperature: Ambient
MDL Number: MFCD35198923
CAS Number: 3002553-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR