1-(2,4-Difluoro-3-(methylthio)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

380466-5G 380466-25G 380466-1G 380466-500MG
77628-978EA 1484.23 USD
77628-978 77655-516 77642-506 77646-522
1-(2,4-Difluoro-3-(methylthio)phenyl)propan-1-one ≥95%
1-(2,4-Difluoro-3-(methylthio)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD35198917
CAS Number: 3002516-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR