4-(9H-carbazol-9-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

38039-5G 38039-25G
77634-958EA 160 USD
77634-958 77655-514
4-(9H-carbazol-9-yl)phenol ≥97%
Formula: C₁₈H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD23111176
CAS Number: 222620-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR