1-(2,6-Dichloro-3-(methylthio)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

380388-5G 380388-1G 380388-25G 380388-500MG
77628-976EA 1484.23 USD
77628-976 77638-500 77655-512 77646-520
1-(2,6-Dichloro-3-(methylthio)phenyl)prop-2-en-1-ol ≥95%
1-(2,6-Dichloro-3-(methylthio)phenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 249.157 g/mol
MW: C₁₀H₁₀Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD35198846
CAS Number: 3002535-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR