2-Bromo-1,3-dichloro-4-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

380364-5G 380364-1G 380364-25G 380364-500MG
77628-974EA 1484.23 USD
77628-974 77638-498 77659-086 77650-512
2-Bromo-1,3-dichloro-4-ethylbenzene ≥95%
2-Bromo-1,3-dichloro-4-ethylbenzene

Precaución: For Research Use Only

Formula: C₈H₇BrCl₂
Storage Temperature: Ambient
MDL Number: MFCD35198822
CAS Number: 2168312-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR