1-(2,6-Dichloro-3-ethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

380362-5G 380362-25G 380362-1G 380362-500MG
77628-972EA 1484.23 USD
77628-972 77659-084 77642-504 77646-518
1-(2,6-Dichloro-3-ethylphenyl)ethanol ≥95%
1-(2,6-Dichloro-3-ethylphenyl)ethanol

Precaución: For Research Use Only

Formula: 219.108 g/mol
MW: C₁₀H₁₂Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD35198820
CAS Number: 3002466-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR