(2,6-Dichloro-3-ethylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

380324-5G 380324-25G 380324-1G 380324-500MG
77633-018EA 1441 USD
77633-018 77659-082 77642-502 77646-516
(2,6-Dichloro-3-ethylphenyl)(ethyl)sulfane ≥95%
(2,6-Dichloro-3-ethylphenyl)(ethyl)sulphane

Precaución: For Research Use Only

Formula: 235.173 g/mol
MW: C₁₀H₁₂Cl₂S
Storage Temperature: Ambient
MDL Number: MFCD35198782
CAS Number: 3002449-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR