(2,6-Dichloro-3-ethylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

380321-5G 380321-25G 380321-1G 380321-500MG
77633-016EA 1441 USD
77633-016 77655-510 77642-500 77646-514
(2,6-Dichloro-3-ethylphenyl)trimethylsilane ≥95%
(2,6-Dichloro-3-ethylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 247.236 g/mol
MW: C₁₁H₁₆Cl₂Si
Storage Temperature: Ambient
MDL Number: MFCD35198779
CAS Number: 3002429-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR