2,2-Difluoro-1-(4-fluoro-2-propoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

380076-1G 380076-5G 380076-500MG
77638-496EA 548 USD
77638-496 77659-080 77650-510
2,2-Difluoro-1-(4-fluoro-2-propoxyphenyl)ethanone ≥97%
2,2-Difluoro-1-(4-fluoro-2-propoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35198590
CAS Number: 3002553-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR