2-Chloro-2,2-difluoro-1-(4-fluoro-2-propoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

380074-5G 380074-1G 380074-25G 380074-500MG
77628-970EA 1484.23 USD
77628-970 77638-494 77659-078 77650-508
2-Chloro-2,2-difluoro-1-(4-fluoro-2-propoxyphenyl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(4-fluoro-2-propoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 266.644 g/mol
MW: C₁₁H₁₀ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35198589
CAS Number: 3002464-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR