1-(4-Fluoro-2-propoxyphenyl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

380064-5G 380064-25G 380064-1G 380064-500MG
77628-968EA 1484.23 USD
77628-968 77659-076 77642-498 77650-506
1-(4-Fluoro-2-propoxyphenyl)propan-1-one ≥97%
1-(4-Fluoro-2-propoxyphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 210.245 g/mol
MW: C₁₂H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD35198579
CAS Number: 3002490-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR