1-(4-Fluoro-2-propoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

380063-5G 380063-1G 380063-25G 380063-500MG
77633-014EA 988 USD
77633-014 77638-492 77655-508 77646-512
1-(4-Fluoro-2-propoxyphenyl)ethanone ≥97%
1-(4-Fluoro-2-propoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD14635234
CAS Number: 1342264-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR