1-(5-Bromo-2,3-difluoro-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

379719-5G 379719-1G 379719-25G 379719-500MG
77628-966EA 1484.23 USD
77628-966 77638-490 77659-074 77646-508
1-(5-Bromo-2,3-difluoro-4-methylphenyl)ethanone ≥95%
1-(5-Bromo-2,3-difluoro-4-methylphenyl)ethanone

Precaución: For Research Use Only

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR