2,4,6-Trichloro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37960-5G 37960-1G 37960-25G 37960-500MG
77634-956EA 352 USD
77634-956 77638-486 77659-070 77650-500
2,4,6-Trichloro-3-methoxybenzoic acid ≥97%
2,4,6-Trichloro-3-methoxybenzoic acid

Precaución: For Research Use Only

Formula: C₈H₅Cl₃O₃
Storage Temperature: Ambient
MDL Number: MFCD33403747
CAS Number: 2624417-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR