(2E)-4-(3-Chloro-4-methoxyphenyl)-4-oxo-2-butenoic acid ≥95%

Supplier: AOBChem USA

Warning

379333-1G 379333-5G 379333-500MG
77638-482EA 594 USD
77638-482 77659-068 77650-498
(2E)-4-(3-Chloro-4-methoxyphenyl)-4-oxo-2-butenoic acid ≥95%
(2E)-4-(3-Chloro-4-methoxyphenyl)-4-oxo-2-butenoic acid

Precaución: For Research Use Only

Formula: 240.64 g/mol
MW: C₁₁H₉ClO₄
Storage Temperature: Ambient
MDL Number: MFCD14536914
CAS Number: 627486-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR