2-Bromo-4-hydroxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-hydroxypyridine-3-carboxylic acid, 2-Bromo-3-carboxy-4-hydroxypyridine

379074-5G 379074-1G 379074-25G
77634-954EA 185 USD
77634-954 77638-480 77659-066
2-Bromo-4-hydroxynicotinic acid ≥97%
2-Bromo-4-hydroxynicotinic acid

Precaución: For Research Use Only

Formula: C₆H₄BrNO₃
MW: 218.005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026106
CAS Number: 1150561-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR