2-Bromo-4-fluoro-5-nitroaniline ≥97%

Supplier: AOBChem USA

Warning

379066-5G 379066-25G 379066-1G 379066-500MG
77634-952EA 335 USD
77634-952 77655-500 77642-492 77650-496
2-Bromo-4-fluoro-5-nitroaniline ≥97%
2-Bromo-4-fluoro-5-nitroaniline

Precaución: For Research Use Only

Formula: C₆H₄BrFN₂O₂
Storage Temperature: Ambient
CAS Number: 346433-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR