(2,3-Dibromo-6-fluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

378850-5G 378850-1G 378850-25G 378850-500MG
77633-012EA 1441 USD
77633-012 77638-478 77659-064 77646-506
(2,3-Dibromo-6-fluorophenyl)triethylsilane ≥95%
(2,3-Dibromo-6-fluorophenyl)triethylsilane

Precaución: For Research Use Only

Formula: C₁₂H₁₇Br₂FSi
MW: 368.155 g/mol
Storage Temperature: Ambient
CAS Number: 3031785-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR