Butyl(2-methyl-3-(trifluoromethyl)phenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

378834-5G 378834-25G 378834-1G 378834-500MG
77628-958EA 1484.23 USD
77628-958 77635-742 77642-490 77650-494
Butyl(2-methyl-3-(trifluoromethyl)phenyl)sulfane ≥97%
Butyl(2-methyl-3-(trifluoromethyl)phenyl)sulphane

Precaución: For Research Use Only

Formula: 248.308 g/mol
MW: C₁₂H₁₅F₃S
Storage Temperature: Ambient
CAS Number: 3031799-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR