Methyl 2-(5-bromo-2-fluoro-3-methylphenyl)-2-oxoacetate ≥95%
Supplier: AOBChem USA
|
Warning
|
378786-5G
378786-25G
378786-1G
378786-500MG
77628-954EA
1484.23
USD
77628-954
77635-740
77642-484
77650-490
Methyl 2-(5-bromo-2-fluoro-3-methylphenyl)-2-oxoacetate ≥95%
Methyl 2-(5-bromo-2-fluoro-3-methylphenyl)-2-oxoacetate
Precaución: For Research Use Only
Formula:
C₁₀H₈BrFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD35198249 CAS Number: 3002514-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...