2,5-Dimethylthioanisole ≥97%

Supplier: AOBChem USA

Warning

378714-5G 378714-25G
77634-948EA 61 USD
77634-948 77655-494
2,5-Dimethylthioanisole ≥97%
2,5-Dimethylthioanisole

Precaución: For Research Use Only

Formula: C₉H₁₂S
MW: 152.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789164
CAS Number: 66623-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR