(4-Fluoro-2-methoxyphenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

377997-5G 377997-25G 377997-1G 377997-500MG
77633-008EA 1441 USD
77633-008 77659-054 77642-482 77646-496
(4-Fluoro-2-methoxyphenyl)(phenyl)methanone ≥95%
(4-Fluoro-2-methoxyphenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 230.234 g/mol
MW: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34661658
CAS Number: 2948315-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR