1-(5-Bromo-2,3,4-trifluorophenyl)-1-cyclopropylethanol ≥95%

Supplier: AOBChem USA

Warning

377990-5G 377990-25G 377990-1G 377990-500MG
77628-946EA 1691.26 USD
77628-946 77659-050 77642-478 77646-492
1-(5-Bromo-2,3,4-trifluorophenyl)-1-cyclopropylethanol ≥95%
1-(5-Bromo-2,3,4-trifluorophenyl)-1-cyclopropylethanol

Precaución: For Research Use Only

Formula: 295.096 g/mol
MW: C₁₁H₁₀BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD35197891
CAS Number: 3002461-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR