2-(5-Bromo-2,3,4-trifluorophenyl)-3,3-dimethylbutan-2-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
377987-5G
377987-25G
377987-1G
377987-500MG
77628-944EA
1707.74
USD
77628-944
77659-048
77642-476
77646-490
2-(5-Bromo-2,3,4-trifluorophenyl)-3,3-dimethylbutan-2-ol ≥95%
2-(5-Bromo-2,3,4-trifluorophenyl)-3,3-dimethylbutan-2-ol
Precaución: For Research Use Only
Formula:
311.138 g/mol MW: C₁₂H₁₄BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD35197889 CAS Number: 3002434-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...