2-(5-Bromo-2,3,4-trifluorophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

377983-1G 377983-500MG 377983-5G
77638-472EA 557 USD
77638-472 77652-738 77655-492
2-(5-Bromo-2,3,4-trifluorophenyl)propan-2-ol ≥97%
2-(5-Bromo-2,3,4-trifluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 269.058 g/mol
MW: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD35197885
CAS Number: 1782792-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR