(5-Bromo-2,3,4-trifluorophenyl)(cyclohexyl)methanone ≥97%

Supplier: AOBChem USA

Warning

377975-5G 377975-1G 377975-25G 377975-500MG
77628-942EA 1688.17 USD
77628-942 77638-470 77659-046 77646-488
(5-Bromo-2,3,4-trifluorophenyl)(cyclohexyl)methanone ≥97%
(5-Bromo-2,3,4-trifluorophenyl)(cyclohexyl)methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₂BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD35197877
CAS Number: 3002539-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR